Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Ninjurin-1 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF5105
This item is not returnable.
View return policy
Description
Ninjurin-1 Polyclonal specifically detects Ninjurin-1 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Ninjurin-1 | |
Polyclonal | |
Unconjugated | |
Q92982 | |
NINJ1 | |
E. coli-derived recombinant human Ninjurin-1 Asp2-Val81 Accession # Q92982 | |
100 μg | |
Primary | |
Detects human Ninjurin-1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 5% cross-reactivity with recombinant human Ninjurin-2 is observed. | |
Human | |
IgG |
Western Blot, Immunohistochemistry | |
LYOPH | |
Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
Nerve injury-induced protein 1, nerve injury-induced protein-1, NIN1, NINJ1, NINJURIN, ninjurin 1, ninjurin-1, Ninjurin1 | |
Sheep | |
Affinity Purified | |
RUO | |
4814 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction