Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human NK1R Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB6687
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB6687 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB6687 Supplier R&D Systems Supplier No. MAB6687
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

NK1R Monoclonal specifically detects NK1R in Human samples. It is validated for Western Blot.

Specifications

Antigen NK1R
Applications Western Blot
Classification Monoclonal
Clone 694549
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P25103
Gene Alias NK-1R, NK1Rneurokinin 1 receptor, NKIR, SPR, SPRsubstance-P receptor, TAC1R, TAC1Rtachykinin 1 receptor (substance P receptor, neurokinin 1 receptor), tachykinin receptor 1 (substance P receptor; neurokinin-1 receptor), tachykinin receptor 1NK-1 receptor, TACR1
Gene Symbols TACR1
Host Species Mouse
Immunogen HEK293 human embryonic kidney cell line transfected with human NK1R Accession # P25103
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6869
Test Specificity Detects human NK1R in direct ELISAs.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.