Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human NK1R PE-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FB66871P100
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FB66871P100 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FB66871P100 Supplier R&D Systems Supplier No. FAB66871P100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

NK1R Monoclonal specifically detects NK1R in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen NK1R
Applications Flow Cytometry
Classification Monoclonal
Clone 694501
Conjugate R-PE
Dilution Flow Cytometry 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and sodium azide
Gene Accession No. P25103
Gene Alias NK-1R, NK1Rneurokinin 1 receptor, NKIR, SPR, SPRsubstance-P receptor, TAC1R, TAC1Rtachykinin 1 receptor (substance P receptor, neurokinin 1 receptor), tachykinin receptor 1 (substance P receptor; neurokinin-1 receptor), tachykinin receptor 1NK-1 receptor, TACR1
Host Species Mouse
Immunogen HEK293 human embryonic kidney cell line transfected with human NK1R, Accession No. P25103
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Primary or Secondary Primary
Gene ID (Entrez) 6869.0
Test Specificity Detects human NK1R in ELISAs.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Form Supernatant
Isotype IgG3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.