Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human NKCC1/SLC12A2 Alexa Fluor 750-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB8528S100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB8528S100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB8528S100 Supplier R&D Systems Supplier No. FAB8528S100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

NKCC1/SLC12A2 Monoclonal specifically detects NKCC1/SLC12A2 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen NKCC1/SLC12A2
Applications Flow Cytometry
Classification Monoclonal
Clone 915911
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 750
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. P55011
Gene Alias Basolateral Na-K-Cl symporter, BSC, BSC2, Bumetanide-sensitive sodium-(potassium)-chloride cotransporter 1, NKCC1MGC104233, SLC12A2, solute carrier family 12 (sodium/potassium/chloride transporters), member 2, solute carrier family 12 member 2
Gene Symbols SLC12A2
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human NKCC1/SLC12A2 Met1-Ser1212 Accession # P55011
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6558
Test Specificity Stains NKCC1/SLC12A2 transfectants but not irrelevant transfectants in flow cytometry.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2b
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.