Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human NKG2C/CD159c Alexa Fluor™ 488-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody has been used in 8 publications

$210.00 - $596.50

Specifications

Antigen NKG2C/CD159c/KLRC2
Clone 134591
Dilution Flow Cytometry 5 uL/10^6 cells
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
FAB138G025
GREENER_CHOICE
View Documents
R&D Systems
FAB138G025
25 Tests
Each for $210.00
Only null left
Add to Cart
 
FAB138G100
GREENER_CHOICE
View Documents
R&D Systems
FAB138G100
100 Tests
Each for $596.50
Only null left
Add to Cart
 
Description

Description

NKG2C/CD159c/KLRC2 Monoclonal specifically detects NKG2C/CD159c/KLRC2 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

NKG2C/CD159c/KLRC2
Flow Cytometry 5 uL/10^6 cells
Monoclonal
Purified
RUO
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
3822
BaF3 mouse pro-B cell line transfected with human NKG2C/CD159c and CD94
Primary
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
134591
Flow Cytometry
Alexa Fluor 488
Mouse
Human
CD159 antigen-like family member C, CD159c, CD159c antigen, killer cell lectin-like receptor subfamily C, member 2, KLRC2, MGC138244, NK cell receptor C, NKG2-C, NKG2-C type II integral membrane protein, NKG2CNKG2-C-activating NK receptor
KLRC2
IgG1
Protein A or G purified from hybridoma culture supernatant
Detects human NKG2C/CD159c heterodimer with CD94 in flow cytometry. It does not cross-react with the human NKG2A/CD94 heterodimer or with the human CD94 homodimer.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.