Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human NKG2C/CD159c Alexa Fluor™ 488-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB1381G100UG
This item is not returnable.
View return policy
Description
NKG2C/CD159c/KLRC2 Monoclonal specifically detects NKG2C/CD159c/KLRC2 in Human samples. It is validated for Flow Cytometry.Specifications
NKG2C/CD159c/KLRC2 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
CD159 antigen-like family member C, CD159c, CD159c antigen, killer cell lectin-like receptor subfamily C, member 2, KLRC2, MGC138244, NK cell receptor C, NKG2-C, NKG2-C type II integral membrane protein, NKG2CNKG2-C-activating NK receptor | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
3822 | |
Human | |
Purified |
Flow Cytometry | |
134522 | |
Alexa Fluor 488 | |
Supplied in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
KLRC2 | |
BaF3 mouse pro-B cell line transfected with human NKG2C/CD159c and CD94 | |
100 μg | |
Primary | |
Detects human NKG2C/CD159c. Detects human NKG2C/CD159c as part of the NKG2C/CD94 heterodimer in flow cytometry. No cross-reactivity with the human NKG2A/CD94 heterodimer or with the human CD94 homodimer is detected. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction