Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human NKG2C/CD159c Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 11 publications
$200.50 - $561.50
Specifications
Antigen | NKG2C/CD159c/KLRC2 |
---|---|
Clone | 134522 |
Concentration | LYOPH |
Dilution | Flow Cytometry 0.25 ug/10^6 cells, CyTOF-reported |
Applications | Flow Cytometry, CyTOF |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB1381SP
![]() |
R&D Systems
MAB1381SP |
25 μg |
Each for $200.50
|
|
|||||
MAB1381100
![]() |
R&D Systems
MAB1381100 |
100 μg |
Each for $561.50
|
|
|||||
Description
NKG2C/CD159c/KLRC2 Monoclonal specifically detects NKG2C/CD159c/KLRC2 in Human samples. It is validated for Flow Cytometry, CyTOF-reported.Specifications
NKG2C/CD159c/KLRC2 | |
LYOPH | |
Flow Cytometry, CyTOF | |
Unconjugated | |
Mouse | |
Human | |
CD159 antigen-like family member C, CD159c, CD159c antigen, killer cell lectin-like receptor subfamily C, member 2, KLRC2, MGC138244, NK cell receptor C, NKG2-C, NKG2-C type II integral membrane protein, NKG2CNKG2-C-activating NK receptor | |
KLRC2 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
134522 | |
Flow Cytometry 0.25 ug/10^6 cells, CyTOF-reported | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
3822 | |
BaF3 mouse pro-B cell line transfected with human NKG2C/CD159c and CD94 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human NKG2C/CD159c. Detects human NKG2C/CD159c as part of the NKG2C/CD94 heterodimer in flow cytometry. No cross-reactivity with the human NKG2A/CD94 heterodimer or with the human CD94 homodimer is detected. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title