Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human NKG2D/CD314 Alexa Fluor™ 700-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB1392N100UG
This item is not returnable.
View return policy
Description
NKG2D/CD314 Monoclonal antibody specifically detects NKG2D/CD314 in Human samples. It is validated for Flow CytometrySpecifications
NKG2D/CD314 | |
Monoclonal | |
Alexa Fluor 700 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
CD314, CD314 antigen, D12S2489E, FLJ17759, FLJ75772, Killer cell lectin-like receptor subfamily K member 1, killer cell lectin-like receptor subfamily K, member 1, KLR, KLRK1, NK cell receptor D, NKG2-D, NKG2-D type II integral membrane protein, NKG2-D-activating NK receptor, NKG2DDNA segment on chromosome 12 (unique) 2489 expressed sequence | |
Mouse myeloma cell line NS0-derived recombinant human NKG2D/CD314, Phe78-Val216, Accession # P26718 | |
100 μg | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
1046002 | |
Flow Cytometry | |
P26718 | |
Mouse | |
Protein A or G purified from cell culture supernatant | |
RUO | |
22914 | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction