Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human NKG2D/CD314 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB1392SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1392SP 25 μg
MAB1392100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB1392SP Supplier R&D Systems Supplier No. MAB1392SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

NKG2D/CD314 Monoclonal antibody specifically detects NKG2D/CD314 in Human samples. It is validated for Flow Cytometry

Specifications

Antigen NKG2D/CD314
Applications Flow Cytometry
Classification Monoclonal
Clone 1046002
Conjugate Unconjugated
Dilution Flow Cytometry 0.25 ug/10^6 cells
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS.
Gene Accession No. P26718
Gene Alias CD314, CD314 antigen, D12S2489E, FLJ17759, FLJ75772, Killer cell lectin-like receptor subfamily K member 1, killer cell lectin-like receptor subfamily K, member 1, KLR, KLRK1, NK cell receptor D, NKG2-D, NKG2-D type II integral membrane protein, NKG2-D-activating NK receptor, NKG2DDNA segment on chromosome 12 (unique) 2489 expressed sequence
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human NKG2D/CD314, Phe78-Val216, Accession # P26718
Purification Method Protein A or G purified from cell culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22914
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.