Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human NKp30/NCR3 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB18491SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB18491SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB18491SP Supplier R&D Systems Supplier No. MAB18491SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 7 publications

NKp30/NCR3 Monoclonal specifically detects NKp30/NCR3 in Human samples. It is validated for Agonist Activity.

Specifications

Antigen NKp30/NCR3
Classification Monoclonal
Clone 210847
Concentration LYOPH
Conjugate Unconjugated
Dilution Agonist Activity 1-3 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Alias Activating natural killer receptor p30, CD337, CD337 antigen, CD337activating NK-A1 receptor, LY117,1C7NK-p30, lymphocyte antigen 117, natural cytotoxicity triggering receptor 3, Natural killer cell p30-related protein, NCR3, NKP30, NKp30MALS
Gene Symbols NCR3
Host Species Mouse
Immunogen Mouse T cell hybridoma transfected with human NKp30 and Mouse myeloma cell line NS0-derived recombinant human NKp30 Fc Chimera
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Research Discipline Agonist Activity
Primary or Secondary Primary
Gene ID (Entrez) 259197
Test Specificity Detects human NKp30.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.