Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human NKp30/NCR3 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
Supplier: R&D Systems AF1849
This item is not returnable.
View return policy
Description
NKp30/NCR3 Polyclonal specifically detects NKp30/NCR3 in Human samples. It is validated for Western Blot.Specifications
NKp30/NCR3 | |
Polyclonal | |
Unconjugated | |
Q05D23 | |
NCR3 | |
Mouse myeloma cell line NS0-derived recombinant human NKp30/NCR3 Leu19-Thr138 Accession # Q05D23 | |
100 μg | |
Primary | |
Detects human NKp30/NCR3 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 5% cross-reactivity with recombinant human (rh) NKp80, rhNKp44 and rhNKp46 is observed. | |
Human | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
Activating natural killer receptor p30, CD337, CD337 antigen, CD337activating NK-A1 receptor, LY117,1C7NK-p30, lymphocyte antigen 117, natural cytotoxicity triggering receptor 3, Natural killer cell p30-related protein, NCR3, NKP30, NKp30MALS | |
Goat | |
Affinity Purified | |
RUO | |
259197 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction