Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human NKp30/NCR3 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
$200.50 - $491.00
Specifications
Antigen | NKp30/NCR3 |
---|---|
Clone | 2615A |
Dilution | Flow Cytometry 0.25 ug/10^6 cells, Immunohistochemistry 1-25 ug/mL, CyTOF-ready |
Applications | Flow Cytometry, Immunohistochemistry, CyTOF |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB18492SP
![]() |
R&D Systems
MAB18492SP |
25 μg |
Each for $200.50
|
|
|||||
MAB18492100
![]() |
R&D Systems
MAB18492100 |
100 μg |
Each for $491.00
|
|
|||||
Description
NKp30/NCR3 Monoclonal specifically detects NKp30/NCR3 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, CyTOF-ready.Specifications
NKp30/NCR3 | |
Flow Cytometry 0.25 ug/10^6 cells, Immunohistochemistry 1-25 ug/mL, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
Activating natural killer receptor p30, CD337, CD337 antigen, CD337activating NK-A1 receptor, LY117,1C7NK-p30, lymphocyte antigen 117, natural cytotoxicity triggering receptor 3, Natural killer cell p30-related protein, NCR3, NKP30, NKp30MALS | |
Mouse myeloma cell line NS0-derived human NKp30/NCR3, Leu19-Thr138, Accession # Q05D23 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
2615A | |
Flow Cytometry, Immunohistochemistry, CyTOF | |
Unconjugated | |
Rabbit | |
Human | |
Q05D23 | |
259197 | |
IgG | |
Protein A or G purified from cell culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title