Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human NKp44/NCR2 Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems BAF2249
This item is not returnable.
View return policy
Description
NKp44/NCR2 Polyclonal specifically detects NKp44/NCR2 in Human samples. It is validated for Western Blot.Specifications
NKp44/NCR2 | |
Polyclonal | |
Biotin | |
CAB52289 | |
NCR2 | |
Mouse myeloma cell line NS0-derived recombinant human NKp44/NCR2 Met1-Pro190 Accession # CAB52289 | |
50 μg | |
Primary | |
Detects human NKp44/NCR2 in Western blots. In Western blots, less than 1% cross-reactivity with recombinant human (rh) NKp30, rhNKp46, and rhNKp80 is observed. | |
Human | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
CD336, CD336 antigen, LY95, LY95lymphocyte antigen 95 homolog (activating NK-receptor; NK-p44), lymphocyte antigen 95 (activating NK-receptor; NK-p44), Lymphocyte antigen 95 homolog, natural cytotoxicity triggering receptor 2, Natural killer cell p44-related protein, NCR2, NK cell activating receptor (NKp44), NK cell-activating receptor, NKP44, NK-p44dJ149M18.1 | |
Goat | |
Affinity Purified | |
RUO | |
9436 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction