Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human NKp80/KLRF1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB1900SP
Change view
Click to view available options
Quantity:
25 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1900SP 25 μg
MAB1900 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB1900SP Supplier R&D Systems Supplier No. MAB1900SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody has been used in 2 publications

NKp80/KLRF1 Monoclonal specifically detects NKp80/KLRF1 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.

Specifications

Antigen NKp80/KLRF1
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 239127
Concentration LYOPH
Conjugate Unconjugated
Dilution Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q9NZS2
Gene Alias Activating coreceptor NKp80, CLEC5C, CLEC5CMGC119908, C-type lectin domain family 5 member C, killer cell lectin-like receptor F1, killer cell lectin-like receptor subfamily F member 1, killer cell lectin-like receptor subfamily F, member 1, KLRF1, Lectin-like receptor F1, MGC119907, MGC119909, ML, NKp80
Gene Symbols KLRF1
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant human NKp80/KLRF1 Val66-Tyr231 Accession # Q9NZS2
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51348
Test Specificity Detects human NKp80/KLRF1 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) NKp30, rhNKp44, rhNKp46, or recombinant mouse NKp46 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.