Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human NKp80/KLRF1 APC-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Rat Monoclonal Antibody has been used in 2 publications

$174.60 - $514.50

Specifications

Antigen NKp80/KLRF1
Clone 239127
Dilution Flow Cytometry 10 uL/10^6 cells
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
FAB1900A025
GREENER_CHOICE
View Documents
R&D Systems
FAB1900A025
25 Tests
Each for $174.60
Only null left
Add to Cart
 
FAB1900A100
GREENER_CHOICE
View Documents
R&D Systems
FAB1900A100
100 Tests
Each for $514.50
Only null left
Add to Cart
 
Description

Description

NKp80/KLRF1 Monoclonal specifically detects NKp80/KLRF1 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

NKp80/KLRF1
Flow Cytometry 10 uL/10^6 cells
Monoclonal
Purified
RUO
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Activating coreceptor NKp80, CLEC5C, CLEC5CMGC119908, C-type lectin domain family 5 member C, killer cell lectin-like receptor F1, killer cell lectin-like receptor subfamily F member 1, killer cell lectin-like receptor subfamily F, member 1, KLRF1, Lectin-like receptor F1, MGC119907, MGC119909, ML, NKp80
KLRF1
IgG2a
Protein A or G purified from hybridoma culture supernatant
Detects human NKp80/KLRF1 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) NKp30, rhNKp44, rhNKp46, or recombinant mouse NKp46 is observed.
239127
Flow Cytometry
APC
Rat
Human
Q9NZS2
51348
Mouse myeloma cell line NS0-derived recombinant human NKp80/KLRF1 Val66-Tyr231 Accession # Q9NZS2
Primary
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.