Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human NKp80/KLRF1 C-Terminus Antibody, R&D Systems™

Sheep Polyclonal Antibody
$200.50 - $526.50
Specifications
Antigen | NKp80/KLRF1 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells |
Applications | Western Blot, Flow Cytometry |
Classification | Polyclonal |
Description
NKp80/KLRF1 Polyclonal specifically detects NKp80/KLRF1 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.Specifications
NKp80/KLRF1 | |
Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells | |
Polyclonal | |
Sheep | |
Human | |
Activating coreceptor NKp80, CLEC5C, CLEC5CMGC119908, C-type lectin domain family 5 member C, killer cell lectin-like receptor F1, killer cell lectin-like receptor subfamily F member 1, killer cell lectin-like receptor subfamily F, member 1, KLRF1, Lectin-like receptor F1, MGC119907, MGC119909, ML, NKp80 | |
KLRF1 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot, Flow Cytometry | |
Unconjugated | |
RUO | |
Q9NZS2 | |
51348 | |
Mouse myeloma cell line NS0-derived recombinant human NKp80/KLRF1 C-Terminus Val66-Tyr231 Accession # Q9NZS2 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human NKp80/KLRF1 C-Terminus in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant human (rh) NKp44 and rhNKp30 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title