Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human NKX2.5 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF2444
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF2444 100 μg
AF2444SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF2444 Supplier R&D Systems Supplier No. AF2444
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 4 publications

NKX2.5 Polyclonal specifically detects NKX2.5 in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen NKX2.5
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL
Gene Accession No. P52952
Gene Alias cardiac-specific homeo box, Cardiac-specific homeobox, cardiac-specific homeobox 1, CHNG5, CSX1, CSXFLJ97195, FLJ52202, FLJ99536, Homeobox protein CSX, Homeobox protein NK-2 homolog E, homeobox protein Nkx-2.5, NK2 transcription factor related, locus 5 (Drosophila), NKX2.5FLJ97166, NKX2-5, NKX2E, NKX2EFLJ97197, NKX4-1, tinman paralog
Gene Symbols NKX2-5
Host Species Goat
Immunogen E. coli-derived recombinant human NKX2.5 Gln24-Arg137 Accession # P52952
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1482
Test Specificity Detects human NKX2.5 in direct ELISAs and Western blots.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.