Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human NKX3.1 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF6080
This item is not returnable.
View return policy
Description
NKX3.1 Polyclonal specifically detects NKX3.1 in Human samples. It is validated for Western Blot, Immunocytochemistry.Specifications
NKX3.1 | |
Polyclonal | |
Unconjugated | |
Q99801 | |
NKX3-1 | |
E. coli-derived recombinant human NKX3.1 Met1-Pro123 Accession # Q99801 | |
100 μg | |
Primary | |
Detects human NKX3.1 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) NKX2.2, rhNKX6.1, and rhBAP31 is observed. | |
Human | |
IgG |
Western Blot, Immunocytochemistry | |
LYOPH | |
Western Blot 0.1 ug/mL, Immunocytochemistry 5-15 ug/mL | |
BAPX2, BAPX2homeobox protein Nkx-3.1, Homeobox protein NK-3 homolog A, NK homeobox (Drosophila), family 3, A, NK3 homeobox 1, NK3 transcription factor homolog A, NK3 transcription factor related, locus 1, NK3 transcription factor related, locus 1 (Drosophila), NKX3.1NKX3, NKX3-1, NKX3A, NKX3ANK homeobox, family 3, A | |
Goat | |
Affinity Purified | |
RUO | |
4824 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction