Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human NLRP1/NALP1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB6788SP
This item is not returnable.
View return policy
Description
NLRP1/NALP1 Monoclonal specifically detects NLRP1/NALP1 in Human samples. It is validated for Immunohistochemistry.Specifications
| NLRP1/NALP1 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| CARD7, CARD7SLEV1, Caspase recruitment domain-containing protein 7, CLR17.1, Death effector filament-forming ced-4-like apoptosis protein, DEFCAP, DEFCAPVAMAS1, DKFZp586O1822, KIAA0926DEFCAP-L/S, NACHT, leucine rich repeat and PYD (pyrin domain) containing 1, NACHT, leucine rich repeat and PYD containing 1, NACHT, LRR and PYD containing protein 1, NACHT, LRR and PYD domains-containing protein 1, NACPP1044, NALP1, NALP1caspase recruitment domain protein 7, NLR family, pyrin domain containing 1, Nucleotide-binding domain and caspase recruitment domain, nucleotide-binding oligomerization domain, leucine rich repeat and pyrin domaincontaining 1, PP1044, SLEV1, VAMAS1 | |
| E. coli-derived recombinant human NLRP1/NALP1, Gly1331-Leu1429, Accession # Q9C000 | |
| 25 μg | |
| 22861 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| IgG2b |
| Immunohistochemistry | |
| 447916 | |
| Immunohistochemistry 1-25 μg/mL | |
| Q9C000 | |
| Mouse | |
| Protein A or G purified from cell culture supernatant | |
| Primary | |
| Detects human NLRP1/NALP1 in direct ELISAs. | |
| Human | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction