Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human NLRP3/NALP3 (aa 540-689) Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF7010
This item is not returnable.
View return policy
Description
NLRP3/NALP3 Polyclonal specifically detects NLRP3/NALP3 in Human samples. It is validated for Immunocytochemistry.Specifications
NLRP3/NALP3 | |
Polyclonal | |
Unconjugated | |
Q96P20 | |
NLRP3 | |
E. coli-derived recombinant human NLRP3/NALP3 Lys540-Lys689 Accession # Q96P20 | |
100 μg | |
Primary | |
Detects human NLRP3/NALP3 in direct ELISAs. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) NALP1, rhNALP2, and rhNALP5 is observed. | |
Human | |
IgG |
Immunocytochemistry | |
LYOPH | |
Immunocytochemistry 5-15 ug/mL | |
AGTAVPRL, AII, AII/AVP, Angiotensin/vasopressin receptor AII/AVP-like, AVP, C1orf7, Caterpiller protein 1.1, CIAS1, CLR1.1, Cold autoinflammatory syndrome 1, Cold autoinflammatory syndrome 1 protein, Cryopyrin, Familial cold autoinflammatory syndrome, FCAS, FCU, Leucine-rich repeat-, and PYD-containing protein 3, Muckle-Wells syndrome, MWS, NACHT, NACHT, LRR and PYD containing protein 3, NACHT, LRR and PYD domains-containing protein 3, NALP3, NLR family, pyrin domain containing 3, NLRP3, Nucleotide-Binding Oligomerization Domain, Leucine Rich Repeat And Pyrin Domain Containing 3, PYPAF1, PYRIN-containing APAF1-like protein 1 | |
Goat | |
Affinity Purified | |
RUO | |
114548 | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction