Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human NM23-H1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB6256SP
This item is not returnable.
View return policy
Description
NM23-H1 Monoclonal specifically detects NM23-H1 in Human samples. It is validated for Western Blot.Specifications
NM23-H1 | |
Monoclonal | |
LYOPH | |
Western Blot 0.5 ug/mL | |
P15531 | |
NME1 | |
E. coli-derived recombinant human NM23-H1 Met1-Glu152 Accession # P15531 | |
25 μg | |
Primary | |
Detects human NM23-H1 in Western blots. In Western blots, no cross-reactivity with recombinant human NM23-H2 is observed. | |
Human | |
Purified |
Western Blot | |
642817 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
EC 2.7.4.6, GAAD, Granzyme A-activated DNase, Metastasis inhibition factor nm23, NB, NBS, NDK A, NDKA, NDP kinase A, NDPKA, NDPK-A, NM23A, NM23AWD, nm23-H1, NM23H1, NME1, non-metastatic cells 1, protein (NM23A) expressed in, nucleoside diphosphate kinase A, Tumor metastatic process-associated protein | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
4830 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction