Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human NM23-H2 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF6665SP
This item is not returnable.
View return policy
Description
NM23-H2/NME2 Polyclonal specifically detects NM23-H2/NME2 in Human samples. It is validated for Western Blot, Immunocytochemistry.Specifications
NM23-H2/NME2 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
EC 2.7.13.3, Histidine protein kinase NDKB, MGC2212, NDK B, NDKB, NDP Kinase B2, NDPKB, NDPK-B, NM23B, nm23-H2, NME2, non-metastatic cells 2, protein (NM23B) expressed in, nucleoside diphosphate kinase B, protein (NM23) expressed in | |
Sheep | |
Affinity Purified | |
RUO | |
4831 | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Immunocytochemistry | |
LYOPH | |
Western Blot 1 ug/mL, Immunocytochemistry 5-15 ug/mL | |
P22392 | |
NME2 | |
E. coli-derived recombinant human NM23-H2 Ala2-Glu152 Accession # P22392 | |
25 μg | |
Primary | |
Detects human NM23-H2 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human NM23-H1 is observed. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction