Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human nNOS Alexa Fluor 700-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. IC24161N100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC24161N100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC24161N100 Supplier R&D Systems Supplier No. IC24161N100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

nNOS Monoclonal specifically detects nNOS in Human samples. It is validated for Intracellular Staining by Flow Cytometry.

Specifications

Antigen nNOS
Applications Flow Cytometry
Classification Monoclonal
Clone 85340
Conjugate Alexa Fluor 700
Dilution Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Gene Accession No. P29475
Gene Alias bNOS, brain, Constitutive NOS, EC 1.14.13.39, IHPS1, Neuronal NOS, nitric oxide synthase 1 (neuronal), N-NOS, nNOSNOS type I, NOS, NOS1, Peptidyl-cysteine S-nitrosylase NOS1
Host Species Mouse
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human nNOS, Ser218-Ser1434, Accession # P29475
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4842
Test Specificity Detects human nNos in direct ELISAs and Western blots.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.