Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Nogo-C Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF5766SP
This item is not returnable.
View return policy
Description
Nogo-C Polyclonal specifically detects Nogo-C in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Nogo-C | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Sheep | |
Affinity Purified | |
RUO | |
57142 | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Immunohistochemistry | |
LYOPH | |
Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
RTN4 | |
11 aa unique to the N-terminus of human Nogo-C synthetic peptide | |
25 μg | |
Primary | |
Detects Nogo-C in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) Nogo-A and rhNogo-B is observed. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction