Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Nogo Receptor/NgR Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB1208
This item is not returnable.
View return policy
Description
Nogo Receptor/NgR Monoclonal specifically detects Nogo Receptor/NgR in Human samples. It is validated for Western Blot.Specifications
Nogo Receptor/NgR | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL | |
Q9BZR6 | |
RTN4R | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Nogo Receptor/NgR Cys27-Ser447 (predicted) Accession # Q9BZR6 | |
500 μg | |
Primary | |
Detects human Nogo Receptor/NgR in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse Nogo Receptor/NgR is observed. | |
Human | |
Purified |
Western Blot | |
188428 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
NgR, NgR1, Nogo receptor, Nogo-66 receptor, NogoR, NOGORUNQ330/PRO526, reticulon 4 receptor, reticulon-4 receptor, RTN4R | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
65078 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction