Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Norrin Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 4 publications
$200.50 - $549.50
Specifications
Antigen | Norrin/NDP |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Blockade of Receptor-ligand Interaction 2-6 ug/mL |
Applications | Western Blot, Immunohistochemistry, Blockade of Receptor-ligand Interaction |
Classification | Polyclonal |
Description
Norrin/NDP Polyclonal specifically detects Norrin/NDP in Human samples. It is validated for Western Blot, Immunohistochemistry, Blockade of Receptor-ligand Interaction.Specifications
Norrin/NDP | |
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Blockade of Receptor-ligand Interaction 2-6 ug/mL | |
Polyclonal | |
Goat | |
Human | |
EVR2, exudative vitreoretinopathy 2 (X-linked), FEVR, ND, NDP, Norrie disease (pseudoglioma), Norrie disease protein, norrin, X-linked exudative vitreoretinopathy 2 protein | |
NDP | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot, Immunohistochemistry, Blockade of Receptor-ligand Interaction | |
Unconjugated | |
RUO | |
Q00604 | |
4693 | |
E. coli-derived recombinant human Norrin Lys25-Ser133 Accession # Q00604 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human Norrin in direct ELISAs and Western blots. In direct ELISAs, approximately 50% cross-reactivity with recombinant mouse Norrin is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title