Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Notch-1 Alexa Fluor 750-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB5317S100UG
This item is not returnable.
View return policy
Description
Notch-1 Monoclonal specifically detects Notch-1 in Human samples. It is validated for Flow Cytometry.Specifications
Notch-1 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
P46531 | |
NOTCH1 | |
Mouse myeloma cell line NS0-derived recombinant human Notch-1 aa 19-526 Accession # P46531 | |
RUO | |
4851 | |
Human | |
IgG1 |
Flow Cytometry | |
527425 | |
Alexa Fluor 750 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
EC 2.1.2.11, EC 3.4.21.68, hN1, Notch (Drosophila) homolog 1 (translocation-associated), notch 1Notch homolog 1, translocation-associated (Drosophila), Notch homolog 1, translocation-associated, Notch1, TAN1, TAN1neurogenic locus notch homolog protein 1, Translocation-associated notch protein TAN-1 | |
Mouse | |
100 μg | |
Primary | |
Detects human Notch-1. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction