Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Notch-1 PE-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 4 publications
$210.00 - $456.00
Specifications
Antigen | Notch-1 |
---|---|
Clone | 527425 |
Dilution | Flow Cytometry 10 uL/10^6 cells |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FAB5317P025
![]() |
R&D Systems
FAB5317P025 |
25 Tests |
Each for $210.00
|
|
|||||
FAB5317P100
![]() |
R&D Systems
FAB5317P100 |
100 Tests |
Each for $456.00
|
|
|||||
Description
Notch-1 Monoclonal specifically detects Notch-1 in Human samples. It is validated for Flow Cytometry.Specifications
Notch-1 | |
Flow Cytometry 10 uL/10^6 cells | |
Monoclonal | |
Purified | |
RUO | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
EC 2.1.2.11, EC 3.4.21.68, hN1, Notch (Drosophila) homolog 1 (translocation-associated), notch 1Notch homolog 1, translocation-associated (Drosophila), Notch homolog 1, translocation-associated, Notch1, TAN1, TAN1neurogenic locus notch homolog protein 1, Translocation-associated notch protein TAN-1 | |
NOTCH1 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Detects human Notch-1. |
527425 | |
Flow Cytometry | |
R-PE | |
Mouse | |
Human | |
P46531 | |
4851 | |
Mouse myeloma cell line NS0-derived recombinant human Notch-1 Ala19-Gln526 Accession # P46531 | |
Primary | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title