Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Notch-3 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB1559SP
This item is not returnable.
View return policy
Description
Notch-3 Monoclonal specifically detects Notch-3 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.Specifications
Notch-3 | |
Monoclonal | |
LYOPH | |
Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
Q9UM47 | |
NOTCH3 | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Notch-3 Ala40-Glu467 Accession # Q9UM47 | |
25 μg | |
Primary | |
Detects human Notch-3 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) Notch-3 ICD (aa 2195-2321), rhNotch-1, rhNotch-1 ICD (aa 2251-2556), rhNotch-2, rhNotch-2 ICD (aa 2063-2413), rhNotch-4, rhNotch-4 ICD (aa 1778-2003), recombinant mouse Notch-3, or recombinant rat DLL1 is observed. | |
Human | |
Purified |
Flow Cytometry, CyTOF | |
603532 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CADASIL, CASILneurogenic locus notch homolog protein 3, Notch (Drosophila) homolog 3, notch 3Notch homolog 3 (Drosophila), Notch homolog 3, Notch3 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
4854 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction