Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Notch-3 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF1559
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF1559 100 μg
AF1559SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF1559 Supplier R&D Systems Supplier No. AF1559
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody has been used in 2 publications

Notch-3 Polyclonal specifically detects Notch-3 in Human samples. It is validated for Western Blot, Blockade of Receptor-ligand Interaction.

Specifications

Antigen Notch-3
Applications Western Blot, Blockade of Receptor-ligand Interaction
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Blockade of Receptor-ligand Interaction
Gene Accession No. Q9UM47
Gene Alias CADASIL, CASILneurogenic locus notch homolog protein 3, Notch (Drosophila) homolog 3, notch 3Notch homolog 3 (Drosophila), Notch homolog 3, Notch3
Gene Symbols NOTCH3
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant human Notch-3 Ala40-Glu467 Accession # Q9UM47
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4854
Test Specificity Detects human Notch-3 in direct ELISAs and Western blots. In direct ELISAs, approximately 50% cross-reactivity with recombinant mouse Notch-3 is observed and less than 5% cross-reactivity with recombinant human (rh) Notch-1 and rhNotch-2 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.