Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Notch-3 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB1559
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1559 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB1559 Supplier R&D Systems Supplier No. MAB1559
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Notch-3 Monoclonal specifically detects Notch-3 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.

Specifications

Antigen Notch-3
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 603532
Concentration LYOPH
Conjugate Unconjugated
Dilution Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q9UM47
Gene Alias CADASIL, CASILneurogenic locus notch homolog protein 3, Notch (Drosophila) homolog 3, notch 3Notch homolog 3 (Drosophila), Notch homolog 3, Notch3
Gene Symbols NOTCH3
Host Species Mouse
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Notch-3 Ala40-Glu467 Accession # Q9UM47
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4854
Test Specificity Detects human Notch-3 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) Notch-3 ICD (aa 2195-2321), rhNotch-1, rhNotch-1 ICD (aa 2251-2556), rhNotch-2, rhNotch-2 ICD (aa 2063-2413), rhNotch-4, rhNotch-4 ICD (aa 1778-2003), recombinant mouse Notch-3, or recombinant rat DLL1 is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.