Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Notch-3 APC-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB1559A
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB1559A 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB1559A Supplier R&D Systems Supplier No. FAB1559A
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

Notch-3 Polyclonal specifically detects Notch-3 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen Notch-3
Applications Flow Cytometry
Classification Polyclonal
Conjugate APC
Dilution Flow Cytometry 10 uL/10^6 cells
Gene Accession No. Q9UM47
Gene Alias CADASIL, CASILneurogenic locus notch homolog protein 3, Notch (Drosophila) homolog 3, notch 3Notch homolog 3 (Drosophila), Notch homolog 3, Notch3
Gene Symbols NOTCH3
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant human Notch-3 Ala40-Glu467 Accession # Q9UM47
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4854
Test Specificity Detects human Notch-3 in direct ELISAs and Western blots. In direct ELISAs and Western blot, approximately 30% cross-reactivity with recombinant mouse Notch-3 is observed and less than 5% cross-reactivity with recombinant rat Notch-2 is observed.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.