Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Notch-3 Biotinylated Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems BAF1559
This item is not returnable.
View return policy
Description
Notch-3 Polyclonal specifically detects Notch-3 in Human samples. It is validated for Western Blot.Specifications
Notch-3 | |
Polyclonal | |
Biotin | |
Q9UM47 | |
NOTCH3 | |
Mouse myeloma cell line NS0-derived recombinant human Notch-3 Ala40-Glu467 Accession # Q9UM47 | |
50 μg | |
Primary | |
Detects human Notch-3 in Western blots. In Western blots, approximately 40% cross-reactivity with recombinant mouse Notch-3 is observed and less than 5% cross-reactivity with recombinant rat Notch-2 is observed. | |
Human | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
CADASIL, CASILneurogenic locus notch homolog protein 3, Notch (Drosophila) homolog 3, notch 3Notch homolog 3 (Drosophila), Notch homolog 3, Notch3 | |
Sheep | |
Affinity Purified | |
RUO | |
4854 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction