Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Notch-4 Intracellular Domain Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems MAB3847SP
This item is not returnable.
View return policy
Description
Notch-4 Monoclonal specifically detects Notch-4 in Human samples. It is validated for Western Blot, Immunocytochemistry.Specifications
Notch-4 | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL, Immunocytochemistry 8-25 ug/mL | |
Q99466 | |
NOTCH4 | |
E. coli-derived recombinant human Notch-4 Intracellular Domain Gly1778-Lys2003 Accession # Q99466 | |
25 μg | |
Primary | |
Detects human Notch-4 Intracellular Domain in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) Notch-1 Intracellular Domain, rhNotch-2 Intracellular Domain, rhNotch-3, recombinant rat (rr) Notch-1 EC, rrNotch-2 EC, or rrDelta-1 is observed. | |
Human | |
Purified |
Western Blot, Immunocytochemistry | |
411913 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
INT3, neurogenic locus notch homolog protein 4, notch 4, Notch homolog 4, NOTCH3, Notch4 | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
4855 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction