Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Notch-4 Intracellular Domain Antibody, R&D Systems™

Goat Polyclonal Antibody
$200.50 - $468.50
Specifications
Antigen | Notch-4 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL, Immunocytochemistry 5-15 ug/mL |
Applications | Western Blot, Immunocytochemistry |
Classification | Polyclonal |
Description
Notch-4 Polyclonal specifically detects Notch-4 in Human samples. It is validated for Western Blot, Immunocytochemistry.Specifications
Notch-4 | |
Western Blot 0.1 ug/mL, Immunocytochemistry 5-15 ug/mL | |
Polyclonal | |
Goat | |
Human | |
INT3, neurogenic locus notch homolog protein 4, notch 4, Notch homolog 4, NOTCH3, Notch4 | |
NOTCH4 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot, Immunocytochemistry | |
Unconjugated | |
RUO | |
Q99466 | |
4855 | |
E. coli-derived recombinant human Notch-4 Intracellular Domain Gly1778-Lys2003 Accession # Q99466 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human Notch-4 Intracellular Domain in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant human (rh) Notch-1 Intracellular Domain and rhNotch-2 Intracellular Domain is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title