Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Notch-4 Intracellular Domain Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB3847
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB3847 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB3847 Supplier R&D Systems Supplier No. MAB3847
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

Notch-4 Monoclonal specifically detects Notch-4 in Human samples. It is validated for Western Blot, Immunocytochemistry.

Specifications

Antigen Notch-4
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 411913
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunocytochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q99466
Gene Alias INT3, neurogenic locus notch homolog protein 4, notch 4, Notch homolog 4, NOTCH3, Notch4
Gene Symbols NOTCH4
Host Species Rat
Immunogen E. coli-derived recombinant human Notch-4 Intracellular Domain Gly1778-Lys2003 Accession # Q99466
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4855
Test Specificity Detects human Notch-4 Intracellular Domain in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) Notch-1 Intracellular Domain, rhNotch-2 Intracellular Domain, rhNotch-3, recombinant rat (rr) Notch-1 EC, rrNotch-2 EC, or rrDelta-1 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.