Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Notum Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $538.00
Specifications
| Antigen | NOTUM |
|---|---|
| Clone | 962216 |
| Dilution | Western Blot 2 ug/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB9356SP
![]() |
R&D Systems
MAB9356SP |
25 μg |
Each for $200.50
|
|
|||||
MAB9356100
![]() |
R&D Systems
MAB9356100 |
100 μg |
Each for $538.00
|
|
|||||
Description
NOTUM Monoclonal specifically detects NOTUM in Human samples. It is validated for Western Blot.Specifications
| NOTUM | |
| Western Blot 2 ug/mL | |
| Monoclonal | |
| Mouse | |
| Human | |
| Q6BXW9 | |
| 147111 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 962216 | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| EC 3.-, notum pectinacetylesterase homolog (Drosophila), protein notum homolog | |
| Chinese hamster ovary cell line CHO-derived recombinant human Notum Ser81-Thr451 Accession # Q6BXW9 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Notum in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title