Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human NOV/CCN3 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
$200.50 - $1929.00
Specifications
Antigen | NOV/CCN3 |
---|---|
Clone | 209808 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB1640SP
![]() |
R&D Systems
MAB1640SP |
25 μg |
Each for $200.50
|
|
|||||
MAB1640100
![]() |
R&D Systems
MAB1640100 |
100 μg |
Each for $410.00
|
|
|||||
MAB1640500
![]() |
R&D Systems
MAB1640500 |
500 μg |
Each for $1,929.00
|
|
|||||
Description
NOV/CCN3 Monoclonal specifically detects NOV/CCN3 in Human samples. It is validated for Western Blot.Specifications
NOV/CCN3 | |
LYOPH | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
P48745 | |
4856 | |
Mouse myeloma cell line NS0-derived recombinant human NOV/CCN3 Thr32-Met357 Accession # P48745 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human NOV/CCN3 in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody does not cross-react with recombinant mouse NOV, recombinant human (rh) CTGF, or rhWISP-1. |
209808 | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CCN family member 3, CCN3, CCN3IGF-binding protein 9, IBP-9, IGFBP-9, IGFBP9novH, Insulin-like growth factor-binding protein 9, nephroblastoma overexpressed gene, Nephroblastoma-overexpressed gene protein homolog, NOVH, protein NOV homolog | |
NOV | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title