Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Nox4 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF8158
This item is not returnable.
View return policy
Description
Nox4 Polyclonal specifically detects Nox4 in Human samples. It is validated for Western Blot.Specifications
Nox4 | |
Polyclonal | |
Unconjugated | |
Q9NPH5 | |
NOX4 | |
E. coli-derived recombinant human Nox4 Lys450-Ser578 Accession # Q9NPH5 | |
100 μg | |
Primary | |
Detects human Nox4 in direct ELISAs and Western blots. | |
Human | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 2 ug/mL | |
EC 1.6.3, Kidney oxidase-1, Kidney superoxide-producing NADPH oxidase, KOX, KOX-1, KOX-1Renal NAD(P)H-oxidase, NADPH oxidase 4, RENOX, RENOXEC 1.6.3.- | |
Sheep | |
Affinity Purified | |
RUO | |
50507 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction