Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human NPM3 Full-length ORF (NP_008924.1, 1 a.a. - 178 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89954484
Click to view available options
Quantity:
10 μg
25 μg

Used for AP, Array, ELISA, WB-Re

The protein encoded by this gene is related to the nuclear chaperone phosphoproteins, nucleoplasmin and nucleophosmin. This protein is strongly expressed in diverse cell types where it localizes primarily to the nucleus. Based on its similarity to nucleoplasmin and nucleophosmin, this protein likely functions as a molecular chaperone in the cell nucleus. [provided by RefSeq]

Sequence: MAAGTAAALAFLSQESRTRAGGVGGLRVPAPVTMDSFFFGCELSGHTRSFTFKVEEEDDAEHVLALTMLCLTEGAKDECNVVEVVARNHDHQEIAVPVANLKLSCQPMLSLDDFQLQPPVTFRLKSGSGPVRITGRHQIVTMSNDVSEEESEEEEEDSDEEEVELCPILPAKKQGGRP

Specifications

Accession Number NP_008924.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 10360
Molecular Weight (g/mol) 45.7kDa
Name NPM3 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Immunogen MAAGTAAALAFLSQESRTRAGGVGGLRVPAPVTMDSFFFGCELSGHTRSFTFKVEEEDDAEHVLALTMLCLTEGAKDECNVVEVVARNHDHQEIAVPVANLKLSCQPMLSLDDFQLQPPVTFRLKSGSGPVRITGRHQIVTMSNDVSEEESEEEEEDSDEEEVELCPILPAKKQGGRP
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias PORMIN/TMEM123
Common Name NPM3
Gene Symbol NPM3
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.