Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human NPRA/NPR1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB4860
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB4860 100 μg
MAB4860SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB4860 Supplier R&D Systems Supplier No. MAB4860
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

NPRA/NPR1 Monoclonal specifically detects NPRA/NPR1 in Human samples. It is validated for Western Blot.

Specifications

Antigen NPRA/NPR1
Applications Western Blot
Classification Monoclonal
Clone 377029
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P16066
Gene Alias ANPa, ANP-A, ANPRA, ANPR-A, ANPRAEC 4.6.1.2, atrial natriuretic peptide receptor 1, Atrial natriuretic peptide receptor type A, atrionatriuretic peptide receptor A, GC-A, Guanylate cyclase A, GUC2A, GUCY2A, natriuretic peptide A type receptor, natriuretic peptide receptor A, natriuretic peptide receptor A/guanylate cyclase A (atrionatriuretic peptidereceptor A), NPR1, NPRA, NPR-A, Pndr
Gene Symbols NPR1
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human NPRA/NPR1 Gly33-Glu473 Accession # P16066
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4881
Test Specificity Detects human NPRA/NPR1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse NPR1 or recombinant human NPR2 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.