Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human NPRB/NPR2 Alexa Fluor™ 405-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. RD10000085
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RD10000085 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. RD10000085 Supplier R&D Systems Supplier No. FAB9725V100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

NPRB/NPR2 Monoclonal antibody specifically detects NPRB/NPR2 in Human samples. It is validated for Flow Cytometry

Specifications

Antigen NPRB/NPR2
Applications Flow Cytometry
Classification Monoclonal
Clone 1060722
Conjugate Alexa Fluor 405
Dilution Flow Cytometry
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Gene Accession No. P20594
Gene Alias AMD1;AMDM;ANPb;ANPRB;Atrial natriuretic peptide receptor 2;ECDM;GCB;GC-B;GUC2B;GUCY2B;NPRB;NPRBi;SNSK, AMDM, ANPb, ANPRB, ECDM, GC-B, GUC2B, NPR2, NPRBi, SNSK
Host Species Mouse
Immunogen Mouse myeloma cell line, NS0-derived human NPRB/NPR2, Arg23-Ile458, Accession # P20594
Purification Method Protein A or G purified from cell culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4882
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.