Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human NPRC/NPR3 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB10233SP
This item is not returnable.
View return policy
Description
Natriuretic Peptide Receptor C Monoclonal antibody specifically detects Natriuretic Peptide Receptor C in Human samples. It is validated for ImmunohistochemistrySpecifications
Natriuretic Peptide Receptor C | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
ANP-C, ANPR-C, ANPRCFLJ14054, Atrial natriuretic peptide clearance receptor, atrial natriuretic peptide receptor 3, Atrial natriuretic peptide receptor type C, C5orf23, chromosome 5 open reading frame 23, GUCY2B, lgj, natriuretic peptide receptor C/guanylate cyclase C (atrionatriuretic peptidereceptor C), NPR3, NPRC, NPR-C, stri | |
Human embryonic kidney cell, HEK293-derived human NPRC/NPR3, Met1-Glu481, Accession # P17342 | |
25 μg | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. | |
IgG |
Immunohistochemistry | |
1060839 | |
Immunohistochemistry 5-15 ug/mL | |
P17342 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
4883 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction