Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human NPY2R Alexa Fluor™ 405-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB10211V100UG
This item is not returnable.
View return policy
Description
NPY2R Monoclonal specifically detects NPY2R in Human samples. It is validated for Flow Cytometry.Specifications
NPY2R | |
Monoclonal | |
Alexa Fluor 405 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
neuropeptide Y receptor type 2, neuropeptide Y receptor Y2, NPY2-R, NPY-Y2 receptor, Y2 receptor | |
Mouse myeloma cell line NS0-derived transfected with recombinant human NPY2R, Met1-Val381, Accession # P49146 | |
100 μg | |
4887 | |
Human | |
Supernatant |
Flow Cytometry | |
557521 | |
Flow Cytometry 0.25-1 μg/106 cells | |
P49146 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
Primary | |
Detects human NPY2R in direct ELISAs. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction