Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human NPY2R Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB10211SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB10211SP 25 μg
MAB10211100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB10211SP Supplier R&D Systems Supplier No. MAB10211SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

NPY2R Monoclonal specifically detects NPY2R in Human samples. It is validated for Flow Cytometry, CyTOF-ready.

Specifications

Antigen NPY2R
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 557521
Conjugate Unconjugated
Dilution Flow Cytometry 0.25 μg/106 cells, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. P49146
Gene Alias neuropeptide Y receptor type 2, neuropeptide Y receptor Y2, NPY2-R, NPY-Y2 receptor, Y2 receptor
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived transfected with recombinant human NPY2R, Met1-Val381, Accession # P49146
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Primary or Secondary Primary
Gene ID (Entrez) 4887
Test Specificity Detects human NPY2R in direct ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.