Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human NRAGE Alexa Fluor 350-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB3835U100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB3835U100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB3835U100 Supplier R&D Systems Supplier No. FAB3835U100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

NRAGE/MAGED1 Monoclonal specifically detects NRAGE/MAGED1 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen NRAGE/MAGED1
Applications Flow Cytometry
Classification Monoclonal
Clone 353124
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 350
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. Q9Y5V3
Gene Alias DLXIN-1, MAGE tumor antigen CCF, MAGED1, melanoma antigen family D, 1, melanoma-associated antigen D1, Neurotrophin receptor-interacting MAGE homolog, NRAGE, NRAGE MAGE-D1 antigen
Gene Symbols MAGED1
Host Species Mouse
Immunogen E. coli-derived recombinant human NRAGE Met1-Glu778 Accession # Q9Y5V3
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9500
Test Specificity Detects human NRAGE in direct ELISAs.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2b
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.