Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human NRAGE Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $502.50
Specifications
Antigen | NRAGE/MAGED1 |
---|---|
Clone | 353124 |
Concentration | LYOPH |
Dilution | Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 8-25 ug/mL, CyTOF-ready |
Applications | Flow Cytometry, Immunocytochemistry, CyTOF |
Description
NRAGE/MAGED1 Monoclonal specifically detects NRAGE/MAGED1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOF-ready.Specifications
NRAGE/MAGED1 | |
LYOPH | |
Flow Cytometry, Immunocytochemistry, CyTOF | |
Unconjugated | |
Mouse | |
Human | |
Q9Y5V3 | |
9500 | |
E. coli-derived recombinant human NRAGE Met1-Glu778 Accession # Q9Y5V3 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human NRAGE in direct ELISAs. |
353124 | |
Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 8-25 ug/mL, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
DLXIN-1, MAGE tumor antigen CCF, MAGED1, melanoma antigen family D, 1, melanoma-associated antigen D1, Neurotrophin receptor-interacting MAGE homolog, NRAGE, NRAGE MAGE-D1 antigen | |
MAGED1 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title