Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human NRAMP1/SLC11A1 Alexa Fluor 594-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB8400T100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB8400T100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB8400T100 Supplier R&D Systems Supplier No. FAB8400T100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

NRAMP1/SLC11A1 Monoclonal specifically detects NRAMP1/SLC11A1 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen NRAMP1/SLC11A1
Applications Flow Cytometry
Classification Monoclonal
Clone 910825
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 594
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. NP_000569
Gene Alias natural resistance-associated macrophage protein 1, LSH, member 1, NRAMP 1, NRAMP1solute carrier family 11 (sodium/phosphate symporters), member 1, NRAMPnatural resistance-associated macrophage protein 1, SLC11A1, solute carrier family 11 (proton-coupled divalent metal ion transporters)
Gene Symbols SLC11A1
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human NRAMP1/SLC11A1 Met1-Gly550 Accession # NP_000569
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6556
Test Specificity Detects HEK293 human embryonic kidney cell line transfected with human NRAMP1/SLC11A1 by Flow Cytometry. Does not detect untransfected or irrelevant transfected HEK293 cells.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2b
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.