Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human NRAMP1/SLC11A1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB8400
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB8400 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB8400 Supplier R&D Systems Supplier No. MAB8400
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

NRAMP1/SLC11A1 Monoclonal specifically detects NRAMP1/SLC11A1 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.

Specifications

Antigen NRAMP1/SLC11A1
Applications Western Blot, Flow Cytometry, CyTOF
Classification Monoclonal
Clone 910825
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. NP_000569
Gene Alias natural resistance-associated macrophage protein 1, LSH, member 1, NRAMP 1, NRAMP1solute carrier family 11 (sodium/phosphate symporters), member 1, NRAMPnatural resistance-associated macrophage protein 1, SLC11A1, solute carrier family 11 (proton-coupled divalent metal ion transporters)
Gene Symbols SLC11A1
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human NRAMP1/SLC11A1 Met1-Gly550 Accession # NP_000569
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6556
Test Specificity Detects HEK293 human embryonic kidney cell line transfected with human NRAMP1/SLC11A1 by Flow Cytometry. Does not detect untransfected or irrelevant transfected HEK293 cells.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.