Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human NRG1-beta 1/HRG1-beta 1 EGF Domain Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB3771100
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB3771100 100 μg
MAB3771SP 25 μg
MAB3771500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB3771100 Supplier R&D Systems Supplier No. MAB3771100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 2 publications

Neuregulin-1 beta 1/NRG1 beta 1 Monoclonal specifically detects Neuregulin-1 beta 1/NRG1 beta 1 in Human samples. It is validated for ELISA Capture (Matched Antibody Pair).

Specifications

Antigen Neuregulin-1 beta 1/NRG1 beta 1
Applications ELISA
Classification Monoclonal
Clone 147705
Conjugate Unconjugated
Dilution ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. AAA58639
Gene Alias ARIA, GGF, GGF2, Heregulin-1, HGL, HRG, HRG1, HRG1-alpha, HRG1-beta 1, HRGA, MST131, MSTP131, NDF, neuregulin 1, Neuregulin1, Neuregulin1 beta 1, NRG1, NRG1-IT2, SMDF
Gene Symbols NRG1
Host Species Mouse
Immunogen E. coli-derived recombinant human NRG1-beta 1/HRG1-beta 1 Ser2-Lys246 Accession # AAA58639
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3084
Test Specificity Detects human NRG1-beta 1 in ELISAs. Recognizes an epitope found in the EGF-like domain of NRG1-beta 1 (amino acids 179-246). It also detects recombinant human SMDF (Sensory and Motor neuron-Derived Factor), but it does not recognize the EGF-like domain of human NRG1-alpha (amino acids 177-241). The epitope is present within amino acids 213-230 of human NRG1-beta 1. This segment is present in all NRG1 beta isoforms, GGF-2 (glial growth factor), and SMDF, but it is absent from NRG1-alpha , NDF43, and NRG1-gamma .
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.